Would you like to log in again?
Wash Bottles, Color-Coded
Supplier: Heathrow Scientific
HS120247 HS120246 HS120245 HS120248 HS120249
10754-136PK 36.88 USD InStock
10754-136 10754-114 10754-134 10754-112 10754-116
Wash Bottles, Color-Coded
Bottles Wash Bottles
Wash bottles feature a closure and stem that are molded in one piece to ensure leak-proof sealing.
  • Wide bottle mouth makes filling easier
  • Bottle are made from LDPE, with a PP closure
The angled neck and tip provide a uniform stream of liquid, and the tip can be cut back if necessary to increase flow. Color coding (for user, application, or location) enables quick and easy identification.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty