Would you like to log in again?
VWR® Disposable Plastic Transfer Pipette, Graduated
Supplier: VWR
218C.VW
10754-276CS 624.51 USD InStock
10754-276
VWR® Disposable Plastic Transfer Pipette, Graduated
Pipettes Pasteur/Transfer Pipettes
VWR® Disposable Transfer Pipettes are designed to safely transfer and dispense liquids in all types of laboratories.
  • Polyethylene construction
  • Allows for highly controlled liquid dispensation
  • Nontoxic
VWR® pipettes are produced using an optimal blend of low density polyethylene, allowing for highly controlled liquid dispensing procedures.
This VWR® line of pipettes is packaged using medical-grade paper and plastic film. These non-toxic pipettes can be safely incinerated or heat-sealed and frozen at temperatures as low as –70 °C (–94 °F).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty