Would you like to log in again?
Low Ash Quantitative Paper Grade 392, Ahlstrom
Supplier: Ahlstrom
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
3920-4000 3920-0550 3920-1500 3920-0470 3920-0700 3920-2700 3920-0425 3920-2400 3920-1850 3920-1250 3920-0900 3920-3200 3920-1100
10754-458PK 28.52 USD InStock
10754-458 10754-468 10754-474 10754-462 10754-472 10754-460 10754-478 10754-456 10754-466 10754-454 10754-476 10754-464 10754-470
Low Ash Quantitative Paper Grade 392, Ahlstrom
Filters Filter Paper Quantitative Filter Paper
Ahlstrom Grade 392 is a medium rate filtering low ash (Ash 0.01%) filter paper suitable for routine quantitative gravimetric techniques.
It is used for filtration of a wide variety of particles.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty