Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
BAX® System Q7 Startup Package, Hygiena™, Qualicon Diagnostics LLC
Supplier: Qualicon Diagnostics
ASY2016 ASY2020 KIT2026 ASY2018 ESA Q7 MCH2019
10754-680EA 78524.48 USD InStock
10754-680 10754-752 10754-862 10754-678 10754-754 10754-864
BAX® System Q7 Startup Package, Hygiena™, Qualicon Diagnostics LLC
Microbiology Test Systems PCR Tests
The Hygiena™ BAX® System offers advanced, automated detection of foodborne pathogens, spoilage organisms and other microbes in raw ingredients, finished products and environmental samples.
The BAX System uses the power of polymerase chain reaction (PCR) technology to automatically amplify and detect targeted bacterial DNA. First, the DNA is amplified through a series of timed heating and cooling cycles. These cycles produce an exponential increase in the number of targeted DNA fragments, if present, creating millions of copies in a very short time. If the target sequence is present, the BAX System automatically detects the amplified fragments by measuring a fluorescent signal within the sample. If the target sequence is not present, no detectable amplification takes place.
The BAX System Q7 instrument uses optimized PCR assays (sold separately) that target specific DNA fragments unique to each target organism, providing a stable, reliable indicator that the organism is present and vastly reducing the potential for cross reactions with similar organisms. To further simplify the process, the BAX System assays provide all requisite PCR primers, polymerase and nucleotides as a stable, dry, manufactured tablet in sealed PCR tubes to eliminate the need for reagent preparation. After amplification, these tubes remain sealed for the detection phase, significantly reducing the potential for sample or laboratory contamination with the amplified PCR product.
Ordering information: Startup Package includes one BAX System Q7 instrument and computer workstation; two analog heating blocks with inserts and thermometers; three cooling block assemblies; BAX System capping/de-capping tools; multi-channel, adjustable, and repeating pipettes with tips for 96 tests; cluster tube racks with tubes/caps for 96 tests; PCR tube holder tray and racks with tubes/caps for 96 tests; powder-free nitrile gloves; user documentation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty