Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
BAX® System Real-Time PCR Assay for Campylobacter jejuni / coli / lari, Hygiena™, Qualicon Diagnostics LLC
Supplier: Qualicon Diagnostics
KIT2018
10754-888EA 1930.76 USD InStock
10754-888
BAX® System Real-Time PCR Assay for Campylobacter jejuni / coli / lari, Hygiena™, Qualicon Diagnostics LLC
Microbiology Test Systems PCR Tests
The Hygiena™ BAX® System PCR Assay for Campylobacter jejuni/coli/lari produces clear yes-or-no results after a fast processing time in the BAX® System Q7 instrument.
The Hygiena BAX System Real-Time PCR Assay for Campylobacter jejuni/coli/lari produces clear yes-or-no results for all three strains after 70 minutes of processing in the BAX System Q7 instrument. Highly contaminated samples can be tested directly, without enrichment, for even faster results. Certified as an AOAC-RI Performance Tested MethodSM for processed turkey and poultry carcass rinses.
The Hygiena BAX System is an automated detection system that uses the power of the polymerase chain reaction (PCR) to detect harmful bacteria and other microbes in food ingredients, finished products and environmental surfaces.
Ordering information: Includes PCR tubes with tablets, optical caps, protease, and lysis buffer. This assay can only be used with the BAX System Q7 instrument.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty