Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Round Carboy with Spigot, Narrow Mouth
Supplier: VWR
VWR®
583290 583300 584401 584403 584380 584304 584400 584390 583280
10755-130EA 326.18 USD InStock
10755-130 75795-676 75795-698 10755-088 75795-682 10755-116 10755-118 10755-128 10755-090
VWR® Round Carboy with Spigot, Narrow Mouth
Bottles Carboys
Use for storing and dispensing solutions and media. Ideal for sterile water storage.
  • Non sterile
  • Polypropylene, LDPE, and HDPE formats available
  • Excellent chemical resistance
  • Clear graduation marks
  • Narrow-mouth
  • Feature an integrated shoulder handle
Carboys are well suited for use in temperatures ranging from -100 to 100 °C (-148 to 212 °F). Carboys are leak-proof on standard test conditions. Spigots are tied to the handle, and require assembly by hand for usage. No tools are necessary for assembly.
Polypropylene carboys can be autoclaved before refilling to stop recurring bacterial growth. LDPE and HDPE carboys are not autoclavable.
Certifications: Resin meets US FDA 21 CFR, USP Class VI.
Ordering information: Spigot included. Always disassemble spigot and cap from carboy before autoclaving.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty