Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Sterile Flocked Swab, with Molded Plastic Applicator & Nylon Fiber Tip, COPAN Diagnostics
Supplier: COPAN Diagnostics
553C.BX 56380CS01.BX 501CS01.BX 516C.BX 502CS01.BX 560C.BX 511CS01.BX 516CS01.BX 519CS01.BX 520CS01.BX 551C.BX 518CS01.BX 503CS01.BX 520C.BX 528C.BX 552C.BX 525CS01.BX 56780CS01.BX 519C.BX 518C.BX 534CS01.BX 56750CS01.BX
10755-196BX 65.65 USD InStock
10755-196 10755-184 10755-198 10755-220 10755-186 10755-200 10755-222 10755-188 10755-202 10755-224 10755-190 10755-192 10755-180 10755-194 10755-182 10755-204 10755-226 10755-214 10755-206 10755-216 10755-208 10755-218
Sterile Flocked Swab, with Molded Plastic Applicator & Nylon Fiber Tip, COPAN Diagnostics
Swabs Swabs for Sampling
Flocked swabs absorb and immediately release a sample into liquid media
COPAN swabs are compatible with multiple applications and platforms, such as rapid antigen testing, EIA, molecular-based assays, DFA, cytology testing, forensics, bacteriology, and virology culture.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty