Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BRAND Microcentrifuge Tubes and Racks, BrandTech

Supplier: BrandTech

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:


780540 780546 780523 780505 780536 780525 780507 780550 780500 780400
80087-096PK 128.17 USD InStock
80087-096 80087-095 80087-084 80087-083 10215-490 10755-230 10755-232 80087-091 80087-080 80087-078
BRAND Microcentrifuge Tubes and Racks, BrandTech
Tubes Microcentrifuge Tubes
Manufactured from virgin polypropylene, these highly translucent microcentrifuge tubes are free from flash, air bubbles, and streaks.

  • 0.5 ml, 1.5 ml and 2.0 ml sizes available
  • Different colors available
  • Lid closure models withstand up to 30000 RCF

They are produced with uniform wall thickness to consistently withstand centrifugal forces. Attached caps are flat with consistent thickness for trouble-free piercing and form a tight, leak-free seal that is easy to open, even when wearing gloves. Tubes fit most common rotors and may be spun up to 30000 RCF (10000 RCF for 0.5 ml tubes, 20000 RCF for 1.5 ml tubes; 30000 RCF for 1.5 ml with lid closure). BIO-CERT® RNase-, DNase-, and DNA-free tubes are available for critical applications. The 1.5 ml tubes with caps feature graduations. Tubes are colored to simplify sample identification.

Ordering information: BIO-CERT® microcentrifuge tubes are packaged into thirty 15-tube blister packs within the 450 tube overpack.

Order Now
 

ORDER

ACCESSORIES

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty