Would you like to log in again?
BRAND PCR 8- and 12-Strip Tubes and Caps, BrandTech®
Supplier: BrandTech
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Brand®
781334 781280 781324 781413 781330 P1100 781316 781320 781341 781290 781326 P1300 781344 781414 781323 781327 781333 781322 781343 P1200 781321 781332 781340 781317 781342
89093-058PK 123.88 USD InStock
89093-058 10799-765 10799-767 80087-138 89135-434 80087-140 80087-126 10755-236 89400-698 80087-124 80087-122 89093-046 89093-044 89093-042 76183-188 80087-128 10755-234 76183-186 80087-136 76183-184 80087-134 89400-700 80087-132 89093-048 80087-130
BRAND PCR 8- and 12-Strip Tubes and Caps, BrandTech®
Tubes PCR Tubes
BRAND PCR tube strips and strip caps are manufactured from high quality virgin resins without lubricants or chemical additives for reproducible results in PCR applications.
  • Tubes are autoclavable
  • Supplied RNase-, DNase- and DNA-free
New Rigid strips with attached caps for easy handling. Flexible strips for easy strip division.
Strip tubes without attached caps have a tab on the end for easy handling.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty