Would you like to log in again?
Cryogenic Vials, Internal or External Threaded, Simport Scientific
Supplier: Simport
T301-5 T309-2A T308-4A T311-4 T301-4 T310-2A T308-4A T311-1 T310-3A T309-2A T310-5A T310-10A T301-4A T301-2 T301-2 T311-4A T301-1 T310-5A T301-5 T308-2 T311-4A T311-3 T309-3A T309-5A T311-5 T308-3A T301-3 T308-2A T311-2 T301-1 T311-4 T310-2A T309-2 T309-1A T301-4 T309-2 T308-1A T311-5 T309-5A T309-1A T308-5A T301-4A T309-4A T311-3 T309-4A T310-2 T311-1 T301-3 T308-5A T310-1A T309-3A T308-2 T311-2 T308-1A T310-2 T308-3A T308-2A T310-10AES T310-3A T310-4A T310-4A
10755-266CS 866.29 USD InStock
10755-266 CA73960-372 CA73960-370 CA11006-960 CA60872-110 CA73960-376 CA11008-450 CA73960-374 CA60872-150 CA11008-452 CA73960-402 CA11006-956 CA11006-958 CA73960-400 CA11008-448 CA10120-200 CA11006-292 73960-390 89492-662 89108-150 89492-664 60872-130 CA11006-962 89492-660 CA11006-964 CA10120-198 77552-894 89108-148 11006-292 89492-658 89108-146 89108-144 89108-142 89108-140 CA73960-394 89492-650 CA60872-130 CA73960-392 89492-652 CA73960-398 89492-654 CA73960-396 60872-120 89492-656 73960-418 89108-138 89492-648 10062-822 73960-370 CA60872-120 73960-372 73960-378 89492-644 60872-150 89492-646 10062-820 73960-376 CA73960-378 CA73960-418 CA73960-416 CA11028-522
Cryogenic Vials, Internal or External Threaded, Simport Scientific
Vials
Cryogenic Vials are designed for storing biological material, human or animal cells at temperatures as low as −196 °C.
  • Large white marking area and printed graduations
  • Model T310-10A has a polyethylene screw cap
Closures and tubes are both made of polypropylene having the same coefficient of expansion which further enhances the leakproof qualities of these vials at various temperatures.
Certifications: Certified Rnase-, DNase-, Pyrogen- and DNA-free. CE marked.
Ordering information: Tubes can be color coded by the use of CAPINSERT Accessory.
Packaging: Packaged in tamperproof, resealable, safety-lock bags of 100.
Caution: These vials should be used only in the gas phase of liquid nitrogen.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty