Would you like to log in again?
Hydrogen Peroxide WFI Sterile Solutions, 6%
Supplier: Steris
1S0496 1S0508WR 1S0596WR 1S0408 1S0577WR 1S0477 1S3215WR
10755-294CS 774.03 USD InStock
10755-294 10755-296 CA75780-044 10755-298 CA75780-042 CA75780-046 76423-316
Hydrogen Peroxide WFI Sterile Solutions, 6%
Cleaning Agents Detergents and Surface Cleansers
STERIS' 6% Hydrogen Peroxide WFI sterile solution removes residue from hard surfaces even from the most critical surfaces. It is formulated to be a low residue cleaner with minimal organic carbon.
  • Quality - Extremely low level of endotoxins from WFI produced by a Finn-Aqua® Multiple Effect Water Still
  • Ease of Use - Ready to use sterile hydrogen peroxide solution with extremely low odor and residue
  • Convenience - Double bagged for easy introduction into your aseptic areas
  • Safety - 0.2 µm filtered, aseptically filled and packaged minimizes potential for particles being introduces during use
  • Assurance - Each lot documented by a Certificate of Analysis including sterility results
Our 6% Hydrogen Peroxide WFI sterile solution is made with United States Pharmacopeia (USP) quality Water For Injection and Hydrogen Peroxide. It is designed to remove organic and inorganic residue from surfaces. Sterile filtered and filled in an ISO Class 5 aseptic environment, each package is double bagged for easy entry into your controlled environments. This product has an extensive documentation package to meet your validation objectives and is supported by the STERIS Technical Services team for individualized application and validation assistance.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty