Would you like to log in again?
EMS Shears, 7-¹/₄", Emergency Medical International
Supplier: Emergency Medical International
1095-BLUE 1095-BLACK 1095-RED
10755-906EA 12.21 USD InStock
10755-906 10755-908 10755-910
EMS Shears, 7-¹/₄", Emergency Medical International
Scissors
Deluxe EMS Shears features razor sharp, hardened surgical stainless steel blades
Contoured handles for control and comfort. Safety bandage tip. Fully Autoclavable to 143°C. Smooth action yet rugged and durable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty