Would you like to log in again?
Flahback Three™ Light Baton, Emergency Medical International
Supplier: Emergency Medical International
Flashback Three™
3000
10755-990EA 69.07 USD InStock
10755-990
Flahback Three™ Light Baton, Emergency Medical International
Lights Flashlights
The three stage switch provides a flashing light, a steady light and an endcap flashlight
Now available in your choice of red, blue or amber lights. The 20 LED bulbs in each baton can be seen for up to 1 mile and can last up to approximately 90 hours on 3-AA batteries. The Flashback Three™ can be used as a signal light, warning light (with its built in magnetic tailcap) or a dash/deck light. Its weather resistant body is made of high impact ABS plastic and also has a non-slip textured grip with a built-in belt clip and wrist strap. Ideal for all Police, EMS, Fire and 1st Responder personnel.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty