Would you like to log in again?
Flat-Pac™ Mini Kit, Emergency Medical International
Supplier: Emergency Medical International
Flat Pac™
891
10756-146KT 89.64 USD InStock
10756-146
Flat-Pac™ Mini Kit, Emergency Medical International
First Aid Supplies First Aid Kits
The Flat-Pac™ Mini Kit is the most complete miniature first aid kit of its kind.
Downsized from the original Flat-Pac™ it opens completely and lies flat making all contents visible at a glance. One side of the cordura bag has 3 gathered pockets while the other side has one mesh zippered pocket with another open pocket behind it. Its small briefcase style design can easily fit into glove compartments, under car seats, in small storage areas and more. Flat-Pac™ Mini has yellow reflective trim and a Star of Life emblem.
Delivery information: Includes: Bandages and dressings: 2 - 3" sterile conforming bandages, 1 - 5×9" abdominal pad, 4 - 3×3" sterile pads, 1 - triangular bandage, 1 - 1" adhesive tape, 25 - 1×3" adhesive bandages. Airway management: 1 - CPR Lifeshield™ and 1- Plastic bite stick. EMS supplies: 1 - instant ice packs (4×5"), 6- antimicrobial hand wipes, 4 - triple antibiotic ointment, 2 - ammonia inhalants, 2 - latex gloves. Instrument pack: 1 - 5½" bandage scissors, 1 - 4½" splinter forcep, 1 - penlight.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty