Would you like to log in again?
Leica-type Specimen Chucks, Springside Scientific
Supplier: Springside Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SPSD40YL SPSD30PK SPSD40WT SPSD22BL SPSD30WT SPSD55BL SPSD25PK SPSD22WT SPSD25BL SPSD55YL SPSD25WT SPSD40BK SPSD30GN SPSD25OR SPSD30BL SPSD30BK SPSD22RD SPSD40BL SPSD30RD SPSD22GR SPSD25BK SPSD55BK SPSD40PK SPSD30YL SPSD22PK SPSD55GN SPSD55WT SPSD25RD SPSD40RD SPSD25YL SPSD55PK SPSD25GN SPSD22YL SPSD22BK SPSD55RD SPSD40GN SPSD30OR
10756-308EA 73.27 USD InStock
10756-308 10756-208 10756-306 10756-206 10756-304 10756-204 10756-202 10756-292 10756-192 10756-170 10756-290 10756-190 10756-278 10756-212 10756-178 10756-210 10756-176 10756-198 10756-196 10756-174 10756-294 10756-172 10756-194 10756-218 10756-216 10756-214 10756-280 10756-180 10756-168 10756-200 10756-288 10756-166 10756-286 10756-164 10756-284 10756-162 10756-282
Leica-type Specimen Chucks, Springside Scientific
Specimen Mounting Supplies
Suitable for Leica (except for new model CM1950), TBS, Tanner, older Avantik and other common cryostats
  • Orientation mark
  • Proper tolerance and soft aluminum, won't slip in chuck
  • Helps keep track of similar research specimens
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty