Would you like to log in again?
Hareta Replacement Slide Hot Foil Tape, Springside Scientific
Supplier: Springside Scientific
HSFT300
10756-188EA 105.99 USD InStock
10756-188
Hareta Replacement Slide Hot Foil Tape, Springside Scientific
Tapes
This tape is used for automated slide printing and designed for use on Thermo SlideMate printers to replace the expensive tape currently offered by the OEM.
Prints up to 12,000 slides per roll. Will not fade, even in the harshest processing environments, including alcohols and xylene. A direct replacement for the original OEM tape.
Packaging: Each roll is 25 mm x 300 m in length and is a long lasting, standard sized 984' roll.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty