Would you like to log in again?
Leica-Style Cryostat Specimen Chuck Kit, Springside Scientific
Supplier: Springside Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SPGER3010 SPGER2510
10756-298KT 838.94 USD InStock
10756-298 10756-296
Leica-Style Cryostat Specimen Chuck Kit, Springside Scientific
Specimen Mounting Supplies
This frozen section kit is designed for Leica, IEC, Tanner and other common cryostats.
  • Six different o-ring colors
  • Eliminates sample errors and use of expensive colored freezing medium
  • Orientation mark to assure proper sample positioning
  • Proper dimensional tolerance
  • Soft aluminum to avoid slippage slip in chuck that results in thin/thick cutting
  • High-quality angular brush and teasing needle included in kit
  • Quality real wood box
Ordering information: Each kit includes 10 specimen chucks in different o-ring colors and three different sizes. Kit #01 includes six 25 mm chucks, two 30 mm chucks, two 40 mm chucks, an angular brush and a teasing needle. Kit #02 includes six 30 mm chucks, two 25 mm chucks, two 40 mm chucks, an angular brush and a teasing needle.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty