Would you like to log in again?
Zombie NIR™ Fixable Viability Kit, BioLegend
Supplier: BioLegend
423106 423105
10761-294EA 459.03 USD InStock
10761-294 10761-492
Zombie NIR™ Fixable Viability Kit, BioLegend
Assays Cellular Assays
Zombie NIR™ Fixable Viability kit is used in flow cytometric analysis.
  • Composed of lyophilized Zombie NIR™ dye and anhydrous DMSO
  • Analysis of intracellularly-stained cells and antibody surface-stained cells
Zombie NIR™ Fixable Viability kit is composed of lyophilized Zombie NIR™ dye and anhydrous DMSO and is used in flow cytometric analysis of intracellularly-stained cells and antibody surface-stained cells.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty