Would you like to log in again?
Fixable Viability Kit, Zombie Violet™
Supplier: BioLegend
423113 423113 423114 423114
10761-308EA 113.66 USD InStock
10761-308 CA10761-308 CA10761-310 10761-310
Fixable Viability Kit, Zombie Violet™
Assays Cellular Assays
Zombie Violet™ is an amine-reactive fluorescent dye that is non-permeant to live cells but permeant to the cells with compromised membranes
Zombie Violet™ dye is excited by the violet laser (405 nm) and has a fluorescence emission maximum of 423 nm. If using in a multi-color panel design, filter optimization may be required depending on other fluorophores used.
Thus, it can be used to assess live vs. dead status of mammalian cells. Zombie Violet™ is a polar, water soluble dye providing green fluorescence, making it suitable for multi-color detection.
Delivery information: Zombie Violet™ Fixable Viability Kit is composed of lyophilized Zombie Violet™ dye and anhydrous DMSO.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty