Would you like to log in again?
Pacific Blue™ Annexin V Apoptosis Detection Kit, Biolegend®
Supplier: BioLegend
640928 640922 640928 640922
10763-252EA 433.23 USD InStock
10763-252 CA10763-252 CA10763-246 10763-246
Pacific Blue™ Annexin V Apoptosis Detection Kit, Biolegend®
Assays Cellular Assays
Annexin V Apoptosis detection kit has been specifically designed for the identification of apoptotic and necrotic cells.
  • Binding Buffer is recommended for use with Annexin V staining
  • Target and identify apoptotic cells
  • Intracellular proteins that binds to phosphatidylserine (PS) in a calcium-dependent manner
Annexin V (or Annexin A5) is a member of the annexin family of intracellular proteins that binds to phosphatidylserine (PS) in a calcium-dependent manner
Annexin V Binding Buffer is recommended for use with Annexin V staining. Annexin V binding alone cannot differentiate between apoptotic and necrotic cells. To help distinguish between the necrotic and apoptotic cells we recommend use of our Propidium Iodide Solution (PI). Early apoptotic cells will exclude PI, while late stage apoptotic cells and necrotic cells will stain positively, due to the passage of these dyes into the nucleus where they bind to DNA.
Propidium iodide is a fluorescent dye that binds to DNA. When excited by 488 nm laser light, it can be detected with in the PE/Texas Red® channel with a bandpass filter 610/10. It is commonly used in evaluation of cell viability or DNA content in cell cycle analysis by flow cytometry. 7-AAD (7-amino-actinomycin D) has a high DNA binding constant and is efficiently excluded by intact cells. It is useful for DNA analysis and dead cell discrimination during flow cytometric analysis. When excited by 488 laser light, 7-AAD fluorescence is detected in the far red range of the spectrum (650 nm long-pass filter).
PS is normally only found on the intracellular leaflet of the plasma membrane in healthy cells, but during early apoptosis, membrane asymmetry is lost and PS translocates to the external leaflet. Fluorochrome-labeled Annexin V can then be used to specifically target and identify apoptotic cells.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty