Would you like to log in again?
AVA SOFTWARE LEVEL 1
Supplier: Heidolph NA, LLC
Ava Software Level 1^ Kit Includes 1 Year Ava Care. For hotplate users who want to run, record and repeat experiments. Control a single hotplate or up to 4 in parallel. Track and Repeat- create and record simple or complex profiles to control your hotplate.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
AVA SOFTWARE LEVEL 1 10764-054 015200001 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty