Would you like to log in again?
Calcium Standard Solution, 1000 mg/L as CaCO₃, 1 L
Supplier: Hach
12153 12153
10765-188EA 96.7 USD InStock
CA121-53 10765-188
Calcium Standard Solution, 1000 mg/L as CaCO₃, 1 L
Standards Cationic Standards Calcium Standards
For accuracy checks in calcium hardness determinations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty