Would you like to log in again?
Target DP™ Glass Screw-Thread Vials, 9-42, Thermo Scientific
Supplier: Thermo Scientific
Target DP™
C4000-S1W C4000-V2 C5000-680 C4000-LV1 C4000-1 C4000-S1 C4000-LV2 C4000-LV2W C4000-S2W C5000-2W 60180-698 C4000-1W C4000-LV1W 60180-694 C4000-9A C4000-1B C4000-LV3W 60180-693 C4000-9 C4000-9TR C4000-2W C4000-V1 C4000-S9
66030-998PK 416.45 USD InStock
66030-998 10781-888 89235-910 66064-952 10766-770 66030-992 66064-910 66030-002 66030-000 66030-990 66064-912 66064-914 66030-996 66030-006 66064-858 66030-994 82428-552 89187-282 66031-045 97054-234 89235-918 97054-232 97054-242
Target DP™ Glass Screw-Thread Vials, 9-42, Thermo Scientific
Vials
Nearly eliminate septa push-through and improve sealing with Advanced Vial Closure System technology. This innovation not only prevents septa push-through, but provides a stop point for the cap to prevent over - or under-tightening and provide an optimal, consistent seal every time. Vials are made from superior quality 33 expansion borosilicate clear Type 1, Class A glass, and meet all US, EU, and JPN Pharmacopia standards.
  • Closure system prevents septa push-through
  • Wide neck opening
Fill vials easily with the wide neck opening; requires 6 mm diameter micro-inserts.
Thermo Scientific National Preassembled Target DP Kits include 100 2 ml vials with pre-attached caps and septa. Packaged in convenient vial trays with clear covers or in polybags.
Get maximum sample extraction without the need for separate inserts by using Microsampling and High Recovery Vials. Identify sample and volume with graduated, write-on patch provided on certain vials. Obtain maximum recovery of critical polar, labile or OH-interacting compounds using silanized (deactivated) glass (optional). The polyspring microseal inserts self-align and provide a cushion against needle contact. Precision point inserts minimize residual sample loss. Pulled point inserts provide an economical choice for noncritical applications. Vials provide the sealing and performance of a crimp top vial. The Sure Stop vial design incorporates a definite stopping point in the vial finish and removes all subjectivity in achieving optimal seal compression closing a vial. Prevents over- or under-tightening of closures and gives an optimal seal every time. Recommended for most brands of autosampler.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty