Would you like to log in again?
TransIT®-Jurkat Transfection Reagent, Mirus Bio
Supplier: Mirus Bio
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
TransIT®
MIR2125 MIR2126 MIR2124 MIR2122 MIR2120
10766-794PK 2202.97 USD InStock
10766-794 10767-058 10766-790 10766-792 10766-788
TransIT®-Jurkat Transfection Reagent, Mirus Bio
Transfection Reagents
A high-efficiency DNA transfection reagent optimized for Jurkat and other hard-to-transfect cell types.
  • Unique formulation
  • High-efficiency delivery
  • Deliver single or multiple plasmids
Maximize transfection performance in Jurkat cells and other hard-to-transfect cells such as K562, RAW 264.7, and THP-1 cells.
Reagent is suitable for many applications such as gene expression and promoter analysis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty