Would you like to log in again?
TransIT®-Keratinocyte Transfection Reagent, Mirus Bio
Supplier: Mirus Bio
TransIT®
MIR2806 MIR2800 MIR2805 MIR2804
10766-816PK 3822.56 USD InStock
10766-816 10766-812 10766-814 10766-810
TransIT®-Keratinocyte Transfection Reagent, Mirus Bio
Transfection Reagents
A high-efficiency, low-toxicity DNA transfection reagent optimized for keratinocyte cells.
  • Unique formulation
  • Low cellular toxicity
  • High-efficiency delivery
Maximize transfection performance in keratinocyte cells. Achieve 20-30% transfection efficiency with this reagent, ensuring experimental success while maintaining cell density and reducing experimental biases.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty