Would you like to log in again?
TransIT®-Insect Transfection Reagent, Mirus Bio
Supplier: Mirus Bio
TransIT®
MIR6104 MIR6106 MIR6100 MIR6105
10767-032EA 2913.12 USD InStock
10767-032 10766-894 10766-896 10766-898
TransIT®-Insect Transfection Reagent, Mirus Bio
Transfection Reagents
TransIT® -Insect Transfection Reagent is a novel transfection formulation specifically developed for high-performance transfection of plasmid DNA into insect cells such as: Sf9, High Five™ and S2.
  • Exceptional DNA Delivery – In insect cell types including Sf9, High Five™ and S2
  • High Baculovirus Production – Ideal for baculovirus expression in insect cells
  • Serum compatibility
TransIT®-Insect Transfection Reagent is in 80% ethanol and should be stored tightly capped at –20®C.
TransIT-Insect can also be used to transfect insect cells with baculovirus DNA to make recombinant virus. TransIT-Insect is composed of animal-origin free components and is serum compatible, which eliminates the need for any culture medium change after transfection.
Ordering information: TransIT® -Insect Transfection reagent is available in 0.4 mL, 1 mL, 5×1 ml (5-pack) and 10×1 ml (10-pack) configurations.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty