Would you like to log in again?
Label IT® siRNA Tracker Intracellular Localization Kits, Mirus Bio
Supplier: Mirus Bio
MIR7215 MIR7213 MIR7212 MIR7216 MIR7214 MIR7217
10766-926EA 549.53 USD InStock
10766-926 10766-928 10766-934 10766-936 10766-930 10766-932
Label IT® siRNA Tracker Intracellular Localization Kits, Mirus Bio
Assays Nucleic Acid Assays
These Label IT® siRNA Tracker Intracellular Localization Kits are efficient, direct labeling for in vitro and in vivo tracking of siRNA.
  • Labels 50 µg siRNA
  • Easily and consistently control the labeling reactions and labeling density
  • High efficiency labeling with optimal visualization of siRNA in cells and tissues
Generate a labeled siRNA sample without hindering its inherent ability to silence targeted genes. Monitor both sub-cellular localization and functional inhibition of target gene expression simultaneously following introduction of the labeled siRNA into mammalian cells. Contain sufficient reagents to label 50 µg of siRNA. siRNA can be delivered to cells in vitro or in vivo using the transfection reagent of your choice.
Versatile Labeling – Efficiently label and visualize the siRNA of your choice.
One-step chemical method – Easily and consistently control the labeling reactions and labeling density.
High efficiency labeling – Optimal visualization of siRNA in cells and tissues.
Delivery information: Each Kit contains Label IT® siRNA Tracker Reagent, Label IT® Reconstitution Solution, 10X Labeling Buffer A, and siRNA Dilution Buffer.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty