Would you like to log in again?
Label IT® RNAi Delivery Controls, Mirus Bio
Supplier: Mirus Bio
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MIR7902 MIR7901 MIR7900 MIR7903
10766-938EA 195.65 USD InStock
10766-938 10766-944 10766-940 10766-942
Label IT® RNAi Delivery Controls, Mirus Bio
Nucleic Acid Reagents RNAi
Fluorescently labeled RNAi duplexes ideal for in vitro or in vivo tracking
  • Easily visualize transfected cells and assess delivery efficiency using fluorescent microscopy
  • Not known to target any mammalian genes or cause off-target effects
Suitable for co-delivery experiments with functional siRNA.
Delivery information: Supplied as a ready to use 10 µM stock solution with a 10X RNAi Dilution Buffer.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty