Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Label IT® Nucleic Acid Labeling Kits, Mirus Bio
Supplier: Mirus Bio
MIR4100 MIR3425 MIR3400 MIR3100 MIR3600 MIR3200 MIR3225 MIR3725 MIR3800 MIR3625 MIR3325 MIR3700 MIR3125 MIR3825 MIR3300 MIR4125
10766-980EA 811.44 USD InStock
10766-980 10766-982 10767-076 10767-084 10767-074 10766-828 10766-988 10766-824 10766-978 10766-826 10767-082 10766-984 10766-830 10767-072 10766-986 10767-070
Label IT® Nucleic Acid Labeling Kits, Mirus Bio
Assays Nucleic Acid Assays
Label IT® nucleic acid labeling kits are suitable for direct, non-enzymatic labeling of DNA and RNA.
  • Easily and consistently control the labeling reactions
  • Achieve high sensitivity with optimally labeled DNA and RNA
  • Do not impact hybridisation performance
Permanent, non-destructive modification of nucleic acid residues is ideal for many diverse applications. All the Label IT® reagent kit use the same technology, but each attach different fluorophores with distinct excitation and emission spectra.
Delivery information: Each kit contains a Label IT® reagent, Label IT® reconstitution solution, 10X labeling buffer and solutions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty