Would you like to log in again?
TransIT®-EE Delivery Solution and Starter Kit, Mirus Bio
Supplier: Mirus Bio
TransIT®
MIR5340
10767-012EA 362.7 USD InStock
10767-012
TransIT®-EE Delivery Solution and Starter Kit, Mirus Bio
Transfection Reagents
This solution and starter kit is an "enhanced expression" in vivo delivery solution that delivers DNA to the livers of laboratory mice using hydrodynamic tail vein injection.
  • Naked nucleic acid delivery
  • Versatile platform
Achieve increased levels of gene expression than compared to TransIT®-QR Delivery Solution, TransIT- In Vivo Gene Delivery System, polylysine and PEI methods. This solution is suitable for DNA delivery using hydrodynamic tail vein injection to the mouse strain of your choice.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty