Would you like to log in again?
TransIT-HeLaMONSTER® Transfection Kit, Mirus Bio
Supplier: Mirus Bio
TransIT®
MIR2906 MIR2900 MIR2905 MIR2904
10767-064EA 574.2 USD InStock
10767-064 10767-068 10767-066 10766-820
TransIT-HeLaMONSTER® Transfection Kit, Mirus Bio
Transfection Reagents
A high-efficiency, low-toxicity DNA transfection reagent optimized for HeLa cells.
  • Unique formulation
  • Low cellular toxicity
  • High-efficiency delivery
  • Deliver single or multiple plasmids
Achieve a 50-60% transfection efficiency with the use of these kits.
Each kit contains a base and booster reagent to maximize transfection performance in HeLa cells.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty