Would you like to log in again?
Label IT® Nucleic Acid Modifying Kit, Amine, Mirus Bio
Supplier: Mirus Bio
MIR3900 MIR3925
10767-078EA 963.91 USD InStock
10767-078 10767-080
Label IT® Nucleic Acid Modifying Kit, Amine, Mirus Bio
Assays Nucleic Acid Assays
The Label IT® Nucleic Acid Modifying Kits are suitable for direct, non-enzymatic attachment of functional groups to DNA and RNA.
  • Direct, covalent attachment of amine functional groups to any nucleic acid
  • Easily and consistently control the density of nucleic acid modification
  • Permanent, non-destructive modification of nucleic acids, which can then be conjugated to proteins or peptides, or attached to glass surfaces, or beads or plates
Attach amine functional groups covalently to nucleic acids and control the level of modification. For labeling with hydrazine, amine, or amine-reactive dyes and attachment to activated glass surfaces or hydrazide-coated beads or plates.
Delivery information: Each kit contains Label IT® Reagent, Label IT® Reconstitution Solution, labeling buffer, solutions and G50 spin columns.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty