Would you like to log in again?
Label IT® Plasmid Delivery Controls, Mirus Bio
Supplier: Mirus Bio
Label IT®
MIR7907 MIR7906 MIR7904 MIR7905
10767-088EA 223.64 USD InStock
10767-088 10767-090 10767-094 10767-092
Label IT® Plasmid Delivery Controls, Mirus Bio
Transfection Reagents
These fluorescently labeled plasmids are ideal for in vitro and in vivo tracking.
  • Easily visualize transfected cells and assess delivery efficiency using fluorescent microscopy
  • Suitable for co-delivery experiments with functional plasmids
Sensitive - Easily visualize transfected cells and assess delivery efficiency using fluorescent microscopy.
Compatible - Suitable for co-delivery experiments with functional plasmids.
Ease of use - Supplied as a ready to use 0.5 mg/ml concentration for both in vitro and in vivo tracking studies.
Delivery information: Supplied as a ready to use 0.5 mg/ml concentration for both in vitro and in vivo tracking studies.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty