Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 25, 2024. During this time, our site will be not available. We appreciate your patience.
Bench Scale Jacketed Filter Reactor, Ace Glass Incorporated
Supplier: Ace Glass
6386-45 6386-460 6386-20 6386-115 6386-445 6386-06 6386-12 6386-306 6386-107 6386-316 6386-324 6386-528 6386-40 6386-515 6386-510 6386-524 6386-507 6386-50 6386-28 6386-328 6386-16 6386-455 6386-440 6386-55 6386-110 6386-320 6386-520 6386-24 6386-128 6386-120 6386-60 6386-65 6386-450 6386-312 6386-124 6386-465
10099-104EA 2666.56 USD InStock
10099-104 89054-150 89054-130 89054-152 10767-258 10099-102 10098-822 10767-262 10767-260 10098-826 10767-266 10767-264 10098-824 89054-132 89054-154 89054-134 89054-156 89054-158 89054-136 89054-138 89054-140 10767-766 10767-268 10098-828 10767-764 10767-768 89054-128 10767-272 10767-256 89054-142 89054-144 89054-124 10767-270 89054-146 89054-126 89054-148
Bench Scale Jacketed Filter Reactor, Ace Glass Incorporated
Laboratory Reactor Systems
Jacketed filter reactor systems include all of the necessary components to perform single- or multi-step reactions and filtrations in the same vessel
Pressure version; maximum pressure is 35psig at 100°C. Maximum jacket pressure is 8psig.
Systems with Ace-Thred heads are pressure rated to 35psi at 100°C. System sizes vary from 150mL to 6000mL with 60mm (2.4"), 100mm (4"), or 150mm (6") Duran® flanged heads. Reactor bodies feature ball joint inlets/outlets or Ace-Threds, which can be ordered separately. Stand, shelf, chain clamp, and clamp holder not included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty