Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Air Sampling Blower, Ace Glass Incorporated
Supplier: Ace Glass
7511-500 7511-10
89059-258EA 427.74 USD InStock
89059-258 10767-306
Air Sampling Blower, Ace Glass Incorporated
Manifolds
This shaded pole blower is designed to pull air through an air sampling manifold.
Blower has air delivery of 1.7 m3/min. (60 CFM) at 0.0 meters of water static pressure, which assures a minimum pressure drop across each port in the manifold train. Free air RPM is 3030. Operates on 115 volts, 50/60 cycles.
Ordering information: A 230 volt, 50 cycle version of this unit is available via special order. Contact your Avantor representative for details. For aluminum blower mount (not included), see 89059-254 (25 mm) or 89059-256 (50 mm) series.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty