Would you like to log in again?
Jacketed Kilo Scale Reactor, Ace Glass Incorporated
Supplier: Ace Glass
12845-07 12845-06 12845-02 12845-05 12845-04 12845-15 12845-19 12845-11 12845-03 12845-17 12845-09 12845-13
10768-084EA 88743.84 USD InStock
10768-084 10767-518 10768-086 10767-200 10767-860 10768-088 10768-078 10767-204 10767-202 10768-090 10768-080 10768-082
Jacketed Kilo Scale Reactor, Ace Glass Incorporated
Laboratory Reactor Systems
A jacketed reactor base system that is designed for maximum diversity and ease of use
Each base system comes with the basic starting components required. The motor and accessories should be purchased separately, based upon your specific needs.
Ace Glass has developed a simple base system building platform which allows any reactor system to be customized using catalog and/or custom-designed parts.
Ordering information: Base reactor system includes universal stand with motor mount, jacketed flask, multi-neck head, O-ring/gasket and quick-release clamp for head, cap, O-ring, and quick-release clamp for addition port on head, PTFE Ace-Thred bearing for stir shaft, glass stir shaft, shaft collar and shaft coupling, anchor-style agitator, 45° agitator, flush drain seal valve with O-ring and beaded pipe couplings.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty