Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Motic SMZ-161 Stereo Microscope
Supplier: Motic
1101000300452 1101001901771 1101001402361 1101001400141 1101001401841 1101001401861 1101001901791 1101001901781 1101001402841 1101000300641
10767-530EA 195.94 USD InStock
CA89168-446 CA19000-196 CA89168-448 10767-530 10767-520 CA89168-444 CA19000-666 10767-532 10767-522 10767-528
Motic SMZ-161 Stereo Microscope
Microscopes
Employing the Greenough stereoscopic optical system, the Motic SMZ-161 series of stereo microscopes provides large-scale zoom ratio and the clearest image, with high distortion-free resolution and depth of field for sharp, 3-dimensional images
The Greenough optical system delivers clear and distortion-free images with an improved zoom range of 6:1. Parfocality is ensured while running through the complete magnification range; a click-stop mechanism, implemented in the continuous zoom, enables precise reproduction of magnifications. The extended 110mm working distance in its standard configuration gives sufficient space for sample manipulation, using a convincing 3-dimensional upright image. Convincing optical performance, combined with a small footprint and robust mechanics, makes the SMZ-161 ideal for most applications in education and industry.
Designed with a high eye point and a comfortable viewing angle of 45°, the SMZ-161 enables users to work with prolonged hours of viewing without any discomfort felt in the neck and shoulders.
Ordering information: Microscope is supplied with stands, stage accessories, micrometer reticles, additional illumination, polarizing equipment, Darkfield equipment, Barlow lenses. Microscope is customizable with various accessories. For all available options, please contact your local Avantor sales representative. All Motic products come with a one-year electrical and five-year mechanical warranty.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty