Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
SMZ-171 Stereo Microscope, Motic
Supplier: Motic
1101001402941 1101001402711 1101001402931 1100200600754 1100200600762 1100200600758 1101001402951 1100200600767 1101001402961 1101001402741 1100200600754 1100200600767 1100200600758 1100200600762 1101001402731
10769-600EA 1660.14 USD InStock
CA10769-600 10769-600 CA10769-598 10767-556 10767-534 10767-546 10767-554 10767-538 CA10769-604 10767-548 CA10769-602 10767-552 10767-542 10767-550 10767-540
SMZ-171 Stereo Microscope, Motic
Microscopes
Flexible stereo microscopes for advanced research applications in biomedicine and material science. The 6.7:1 Greenough zoom delivers a standard magnification of 7.5x - 50x with 110 mm working distance in a FOV23mm.
  • Eyepieces feature Widefield N-WF10X/23 mm with diopter adjustment
Delivery information: Supplied with the following: Stands, Stage Accessories, Micrometer Reticles, Additional Illumination, Polarizing Equipment, Darkfield Equipment, Barlow Lenses. Microscope is customizable with various accessories. For all available options, please contact your local Avantor sales representative.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty