Would you like to log in again?
Motic BA310 Polarizing Microscope
Supplier: Motic
1100100402241 1100100402251 1100100402251 1100100402241
10767-564EA 4384.31 USD InStock
CA10769-630 10767-564 10767-566 CA10769-628
Motic BA310 Polarizing Microscope
Microscopes
To meet increasing demands for a dedicated, professional polarizing microscope, Motic introduced the BA310Pol for observation of birefringent samples with efficiency and reliability
The BA310Pol comes standard with Motic’s high quality EC Plan objectives (4x, 10x, 40x S, 60x S) set in a reversed quadruple nosepiece and high-eyepoint N-WF PL10x widefield eyepieces for superb image clarity and field diameter. The hard-coated rotating stage features ball bearings for smooth rotation through 360°, with 1° graduations for quick, precise orientation and reference. The BA310Pol is a versatile system suited for a wide range of petrography, minerology, and material science applications where affordability and ease-of-use are key requirements.
The strain-free CCIS optics, built-up around a sturdy frame with centerable objectives, provide reproducible, high fidelity images. Full polarization is provided through the focusable and centerable Bertrand lens, 360° graduated analyzer for optimum image extinction, and a DIN-compliant compensator slot that accepts all established compensators.
Ordering information: Microscope is supplied with all accessories listed above and micrometer reticles. Contact your Avantor representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty