Would you like to log in again?
Digital Stirring Motor and Controller, Hazmat Series, Ace Glass Incorporated
Supplier: Ace Glass
13555-60 13555-50
10767-864EA 12565.92 USD InStock
10767-864 10767-862
Digital Stirring Motor and Controller, Hazmat Series, Ace Glass Incorporated
Stirrers Stirring Tools
A heavy duty, UL and CSA rated hazardous area motor with digital controller
The motor has variable speeds up to 1750 rpm with a maximum torque of 91.5". The motor output is 230 V 60 Hz, 3 phase. The motor shaft is 5/8". The Digital Microdrive control box has a compact wall mountable design, a six-digit LED display, flat panel control, and is IP66/NEMA 4X rated.
The stir motor meets Class I, Division I, Group D requirements for flammable liquids and gases, and Class II, Division II, Groups F&G for combustible dust.
Delivery information: Available in 115 or 230V models, CE and cULus. The stirrer motor, controller, and gear reducer are sold together.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty