Would you like to log in again?
Right Angle Tee Joint to Tube Compression Adapter, Ace Glass Incorporated
Supplier: Ace Glass
12737-24 12737-30 12737-12 12737-48 12737-64 12737-68 12737-02 12737-54 12737-06 12737-60 12737-38 12737-74 12737-72 12737-04 12737-44 12737-36 12737-62 12737-66 12737-20 12737-22 12737-34 12737-56 12737-52 12737-58 12737-08 12737-16 12737-70 12737-14 12737-26 12737-50 12737-10 12737-28 12737-40 12737-42 12737-46 12737-18 12737-32
10768-040EA 55.54 USD InStock
10768-040 10768-042 10768-020 10768-022 10768-044 10768-046 10768-024 10767-854 10767-510 10767-852 10767-858 10767-856 10768-036 10768-014 10768-038 10768-016 10768-018 10768-050 10767-848 10768-052 10768-030 10767-508 10768-054 10768-032 10768-056 10768-034 10767-502 10767-500 10767-846 10767-506 10767-844 10767-504 10767-498 10767-850 10768-048 10768-026 10768-028
Right Angle Tee Joint to Tube Compression Adapter, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
These glass standard taper and spherical joint adapters feature right angle tee ground glass sidearms.
  • Glass [ST] or spherical joint
  • Right angle tee sidearms
The sidearms are used to make tube compression fitting connections to a wide variety of flexible and rigid tubing using PTFE fittings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty