Would you like to log in again?
Unjacketed Kilo Scale Reactor, Ace Glass Incorporated
Supplier: Ace Glass
12846-23 12846-12 12846-13 12846-14 12846-15 12846-17 12846-21 12846-19
10768-094EA 46765.1 USD InStock
10768-094 10768-096 10768-098 10768-100 10768-102 10768-104 10768-106 10768-092
Unjacketed Kilo Scale Reactor, Ace Glass Incorporated
Laboratory Reactor Systems
An unjacketed reactor base system that is designed for maximum diversity and ease of use
Each base system comes with the basic starting components required. The motor and accessories should be purchased separately, based upon your specific needs.
Ace Glass has developed a simple base system building platform which allows any reactor system to be customized using catalog and/or custom-designed parts.
Ordering information: Base reactor system includes universal stand with motor mount, unjacketed flask, multi-neck head, O-ring/gasket and quick-release clamp for head, cap, O-ring, and quick-release clamp for addition port on head, PTFE Ace-Thred bearing for stir shaft, glass stir shaft, shaft collar and shaft coupling, upper turbine, multi-paddle style lower agitator, flush drain seal valve with O-ring, beaded pipe couplings, aluminum heating mantle, girdle mantle, pilot plant temperature controller, thermocouple, thermocouple extension cord, Ace-Thred adapter for thermocouple, PTFE-clad copper heat exchange cooling coil, and Ace-Thred adapters for coil. 30 L and larger reactors also include a 30"-lb torque wrench and a 1/4" socket.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty