Would you like to log in again?
Smart-Tracker™ Wireless Datalogging Module, Thermo Scientific
Supplier: Thermo Scientific
ST100-102 ST100-100 ST101-100 ST101-200 ST100-200 ST100-101
10768-150EA 271.9 USD InStock
10768-150 10768-152 10768-154 10768-156 10768-158 10768-148
Smart-Tracker™ Wireless Datalogging Module, Thermo Scientific
Data Loggers
Smart-Tracker™ makes temperature monitoring in the cold chain easy. All need is a smart phone to wirelessly check stored temperature readings.
  • Share information via the Cloud anywhere in the world
  • Bluetooth wireless connection for use in many types of container and enclosures
There is no dedicated equipment, no docking stations and no complex computer installations. Download the Smartr-Tracker's mobile app: iPhone or android.
Monitor storage facilities, freezers, refrigerators, heating equipment and packages in transit (subject to network speed and availability).
Certifications: Bluetooth certification: FCC, CE.
Delivery information: Includes module with internal sensor, lithium battery, two colored sensor rings and two side magnetic mount.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty