Would you like to log in again?
Apiezon Stopcock/Ground Glass Joint Greases
Supplier: M & I Materials
Apiezon®
L-00025 N-00025 M-00100 M-00025 H-00025
10769-158EA 239.97 USD InStock
10769-158 10769-156 10769-154 10769-152 10769-160
Apiezon Stopcock/Ground Glass Joint Greases
Lubricants
Silicone-free greases for lubricating stopcocks and movable joints in vacuum systems such as desiccators, rotary evaporators, vacuum traps, and ground glass joints
Apiezon® H, L, M, and N greases consist of distilled, petroleum-based, complex hydrocarbons. Vapor pressure curves are available for each product over its working temperature range.
Within the Apiezon® range there is a grease designed to suit most applications. H grease has thermal conductivity properties and high temperature resistance, preventing it from melting at high temperatures (stiffens above 40°C). L and M greases are particularly good lubricants, with L grease being recommended for ultra high vacuum systems and M in slightly less critical, high vacuum systems. Both products show resistance to irradiation. Structural properties make L and M greases effective as a stationary phase in gas-liquid chromatography. N grease is specifically for use in cryogenic applications, displays a rubbery consistency, and works well as a cushioning lubricant.
Greases demonstrate “gettering” properties (the ability to extract chemical impurities), and hence are well-suited to electronic applications as they remove organic impurities from a metal surface and do not form insulating films.
Ordering information: Packaged in aluminum tubes to prevent contact with the atmosphere and reduce contamination from dust or water.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty