Would you like to log in again?
VWR® Tissue Culture Plate Inserts, Polycarbonate (PC) Membrane, Sterilized, Standard Line
Supplier: VWR
TCS100006 TCS004024 TCS000006 TCS005012 TCS003024 TCS100024 TCS001012 TCS002024 TCS002006 TCS001006 TCS003012 TCS001024 TCS005006 TCS000012 TCS005024 TCS003006 TCS002012 TCS100012 TCS000024
10769-224CS 220.27 USD InStock
10769-224 10769-202 10769-236 10769-200 10769-178 10769-222 10769-234 10769-212 10769-208 10769-190 10769-180 10769-176 10769-198 10769-220 10769-232 10769-188 10769-210 10769-186 10769-230
VWR® Tissue Culture Plate Inserts, Polycarbonate (PC) Membrane, Sterilized, Standard Line
Microplates Insert Plates and Inserts
Tissue Culture Plate Inserts feature a thin, polycarbonate membrane that is available in six pore sizes, ranging from 0.1 to 12.0 μm.
  • Temperature range: –20 to 50 °C (–4 to 122 °F)
  • Polycarbonate membrane construction
  • Available in individual recyclable PET packaging
  • Inserts are not autoclavable
  • Non-pyrogenic
  • Transparent and translucent membrane options are available
All inserts are treated for optimal cell attachment. The polycarbonate plate inserts are sterile and assembled with well plates. They will resist most fixing and staining agents. All plates come with lids.
Certifications: ISO 9001, ISO 13485.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty