Would you like to log in again?
Vileda® Professional Compact Bucket System
Supplier: Vileda Professional
149624 149622 149609
10769-304CS 444.16 USD InStock
10769-304 10769-302 10769-300
Vileda® Professional Compact Bucket System
Buckets
High performance in a compact design
  • Perfect for use with all Roll-O-Matic® Mops (without sieve)
  • Solid and stable polypropylene plastic buckets and bails are 100% autoclavable
  • Unique electro-polished sieve (sold separately) with hole pattern, integrated bucket splash guard, and handle support
The single bucket system includes one six gallon bucket and swivel plastic casters. Fully autoclavable, the casters are fixed to the bucket with a stainless steel pin connection. The double bucket system includes two six gallon buckets and an electro-polished stainless steel chasis that is designed to keep the buckets upright. Stainless steel autoclavable casters are fixed to the chasis with a stainless steel screw/nut connection. The electro-polished sieve is sold separately.
Cleanrooms are decreasing in size, affecting the areas to be cleaned as well as the storage rooms for cleaning equipment. The solution is the new Cleanroom Compact Bucket System concept.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty