Would you like to log in again?
Microflex® N29 Nitrile Exam Gloves
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Microflex®
N295 N292 N291 N290 N294 N291 N290 N293
10769-362CS 225.32 USD InStock
CA10769-368 CA10769-366 CA10769-364 CA10769-362 CA10769-360 CA10769-370 10769-362 10769-360
Microflex® N29 Nitrile Exam Gloves
Gloves
These nitrile gloves are manufactured using a soft and stretchy formulation that offers excellent durability as well as comfort.
  • Powder-free
  • Textured for enhanced gripping power
  • Great tactile sensitivity
A three mil thickness provides enhanced tactile sensitivity.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty