Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Hei-Torque Overhead Stirrers, Heidolph North America
Supplier: Heidolph
036093110 036093050 036093150 036093030 036093090 036093070
10769-724EA 3604.91 USD InStock
10769-724 10769-722 10769-730 10769-720 10769-728 10769-726
Hei-Torque Overhead Stirrers, Heidolph North America
Stirrers Overhead Stirrers
The powerful Hei-Torque benchtop stirrers accomplish the most demanding tasks while providing the highest safety in combination with a unique user interface.
The Hei-Torque Ultimate models are ideal for demanding tasks which have to be reproducible and documentable. The extensive number of the additional features and operation modes allows for a perfect adjusment to individual applications. These units are equipped with a 3.2" digital display for ease of operation, and feature a choice of RS-232 or USB Interface to save all process data in a digital file. These units also come with free software to automize and save all process parameters.
The Hei-Torque Expert models are ideal for standard stirring tasks. They are designed to mix and disperse media that requires non-reproducible results in high-viscosity applications. They are equipped with a 2.4" digital display for ease of operation. Newest motor generation for maximum power at minimum noise level.
The electronic stirrers feature a smooth start operation which prevents spills and splashing media. The speed ramps up slowly until set rpm has been reached.
Stay Bar Size: 13Ø×160L mm
Ordering information: Accessories for these benchtop models are not included.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty