Would you like to log in again?
SYNTHETIC CLEAM ROOM PAPER 115LB 8.5X11
Supplier: VWR International
Paper^ Synthetic Cleanroom paper, VWR* 8. 5 x 11, No. 115, synethic paper is a clay-coated two side substrate with unique physical properties resistant to water, tearing, grease and many chemicals. This paper is ideal for documentation in critical environments. 500 sheets/ case
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
SYNTHETIC CLEAM ROOM PAPER 115LB 8.5X11 10769-790 Case of 500 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty