Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
BD™ Universal Viral Transport System, BD Medical
Supplier: BD Medical
BD™
220531 220528 220220 220220 220529
10769-898PK 458.02 USD InStock
10769-898 10769-896 CA90006-624 90006-624 10769-580
BD™ Universal Viral Transport System, BD Medical
Swabs Transport Swabs
The BD™ Universal Viral Transport System (UVT) is designed to transport viruses, chlamydiae, mycoplasmas, and ureaplasmas at room temperature. It is one of the only systems that may be stored and transported at 2 to 25 °C, all in one formulation.
  • No refrigeration requirements: System does not require refrigerated storage or transport on ice, decreasing the risk of specimen rejection and eliminating messy specimen transport
  • Reduced contamination: System features a unique blend of antimicrobics that target and suppress bacterial and fungal contamination without harming viruses, chlamydiae, mycoplasmas, or ureaplasmas
  • Long-term storage: System helps store samples for long periods by preserving viruses and chlamydiae, when stored frozen with a cryoprotectant
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty